M-System 73VR3100-E-R Paperless Recorder with Optional I/O (max. 8 Ai, 8 Di, 8 Do); 24 VDC
Representative image only.
M-System Mfr # 71VR1-E001-R – Item # EW-49006-45
71VR & 73 VR Series
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Chart TypeDigital
  • Input TypeDC voltage; thermocouple; RTD
  • Max Input (VDC)12
  • Min Input (VDC)-12
  • Max Current Input (DC)20
  • Min Current Input (DC)4
  • Sampling Rate100 milliseconds to 10 minutes (programmable)
  • Display TypeTFT LCD (320 x 240 pixel, 5.5\")
  • AlarmRelay (1)
  • DescriptionPaperless Recorder with Optional I/O (max. 8 Ai, 8 Di, 8 Do); 24 VDC
  • Warranty3 year
3 Year WarrantyConformite EuropeenneIP 65
Key Features
  • 3 year warranty
More About this Item
  • Models 71VR1 is a 1/4 DIN size (96 x 96 mm panel cutout), compact paperless recorder
  • Record, display and set an alarm setpoint for max. 8 Ai 8 Di and 8 Do 3.5-inch TFT color LCD
  • Models 73VR2100 can accept either 2, 4, 6, 8, 10 or 12 points of universal input
  • Models 73VR3100 can accept a wide selection of R3 Series I/O modules compatible with the 73VR3100
  • Up to 4 modules can be mounted at the rear side (max. 64 inputs)
  • One of the I/O modules can be replaced with Network Module (DeviceNet, ModBus RTU, ModBus TCP/IP, PROFIBUS-DP, LONWORKS)
  • 5.5-inch TFT color LCD touch-panel display, Windows CE based
  • Data stored in a CF card
  • Real time trending and recording on the PC Recorder software MSR128 via Ethernet
  • REVIEWS