M-System KS2V3-61-M2/UL DC Input Limit Alarm, Dual; 1-5VDC, 100-240VAC
Representative image only.
M-System Mfr # KS2V3-61-M2/UL – Item # EW-45001-14
Monitor a range of alarm conditions
  • Thumbwheel switch adjustments
  • Only 1.2 inch (29.5mm) wide
  • Front LCD display indicating values in scaled engineering unit range is used to program the module
  • Quad or dual trip
  • Program Lockout function
  • Front-panel configurable
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionDC Input Limit Alarm, Dual; 1-5VDC, 100-240VAC
  • Warranty3 Years
3 Year Warranty
Key Features
  • Thumbwheel switch adjustments
  • Only 1.2 inch (29.5mm) wide
  • Front LCD display indicating values in scaled engineering unit range is used to program the module
  • Quad or dual trip
  • Program Lockout function
  • Front-panel configurable
  • 3 year warranty
More About this Item
–– Compact 1/16 DIN size—ideal for panel surface or DIN rail mount
–– 1 to 5 VDC input (KS2V3)
–– Thermocouple or RTD temperature input (KS2TR2)
–– Front LED display indicating values in scaled engineering unit
–– Time constant for the input filter
–– Alarm relay switching delay timer
–– Power ON delay