M-System M2SP-1A-R Frequency Trnsmtr Dry Contact 4-20 Ma24V Dc

Representative image only.
M-System Mfr # M2SP-1A-R/UL 0-100HZ – Item # EW-68467-47
  • Both Mini-M and Mini-MW use a compact-size, plug-in socket base for quick installation or replacement of module without disturbing wiring
  • 2000 VAC Isolation
  • Standard DIN rail or surface mounting
  • Base socket included with modules
  • CE marking and UL/C-UL Class I Div 2 approval
  • 3 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Input typeDry contact
  • Max Output ch1 (mA)20
  • Min Output ch1 (mA)4
  • Power Supply (VDC)24
  • Max Operating Temperature (° C)55
  • Min Operating Temperature (° C)-5
  • Max Operating Temperature (° F)131
  • Min Operating Temperature (° F)23
  • Height (in)2 63/64
  • Width (in)29/32
  • Depth (in)4 7/8
  • Height (cm)7.6
  • Width (cm)2.3
  • Depth (cm)12.4
  • DescriptionFrequency Trnsmtr Dry Contact 4-20 Ma24V Dc
  • Warranty3 Years
3 Year Warranty

Key Features

  • Both Mini-M and Mini-MW use a compact-size, plug-in socket base for quick installation or replacement of module without disturbing wiring
  • 2000 VAC Isolation
  • Standard DIN rail or surface mounting
  • Base socket included with modules
  • CE marking and UL/C-UL Class I Div 2 approval
  • 3 year warranty

More About this Item

M-System's Mini-M Series Signal Conditioners are designed to accept the largest range of process signal inputs and provide a standard and non-standard DC output.

The Mini-MW Series Signal Splitters provide a second isolated output of independent range, which give you the flexibility to add sophisticated distributed control or MIS monitoring to a local loop, without a worry about mismatched impedance, or the threat of one problem system's output signal impacting the other.

Both Mini-M and Mini-MW use a compact-size, plug-in socket base for quick installation or replacement of module without disturbing wiring.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat