M-System R8-SV2 Series Input Module, -10 to 10 VDC, isolated, 2 channel
Representative image only.
M-System Mfr # R8-SV2 – Item # EW-45004-74
Choose input modules suitable for sensing the system parameters that need to be monitored
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Number of Channels2
  • Input TypeDC current
  • Max Input (VDC)10
  • Min Input (VDC)-10
  • DescriptionSeries Input Module, -10 to 10 VDC, isolated, 2 channel
  • Warranty3 year
3 Year Warranty
Key Features
  • 3 year warranty
More About this Item
Inputs supported include analog, temperature, pulse, and discrete. Analog and discrete outputs are also available to add control or alarm functions. Analog outputs interface with other recording devices or proportional input controllers. Use a discrete output module as an alarm contact triggered by transitions, status changes, or count up conditions for any input. Discrete outputs also control on/off cycling of peripheral machinery.
Choose the recorder with the right combination of communication functions and the required DL8 to R8 connector.