MadgeTech SHOCK101-50 Tilt/Acceleration Data Logger -50/+50G, 7 Day Battery
Representative image only.
MadgeTech Mfr # SHOCK101-50 – Item # EW-18003-00
Shock101, Ultra Shock, and TSR101
  • Anodized Aluminum Enclosure
  • Built in Accelerometers
  • Compact Design
  • Tri-Axial Recording
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Number of Channels3
  • Input TypeForce, shock, tilt, or acceleration (X, Y, Z)
  • Max Pressure50 G of force/tilt/shock
  • Min Pressure-50 G of force/tilt/shock
  • Pressure Resolution0.05 G of force/tilt/shock
  • Memory Size349,525 per axis
  • Battery Life7 days
  • Battery Type1 x 9V battery
  • Length (in)3 1/2
  • Height (in)4 13/32
  • Width (in)1
  • DescriptionTilt/Acceleration Data Logger -50/+50G, 7 Day Battery
  • Warranty1 year
1 Year Warranty
Key Features
  • Anodized Aluminum Enclosure
  • Built in Accelerometers
  • Compact Design
  • Tri-Axial Recording
  • 1 year warranty
More About this Item
The Shock101 is a tri-axial shock recorder that measures and records peak acceleration over a user-specified time period. The time period can be programmed using the MadgeTech software from 64 samplings per second up to 5 minutes. Independent of the user defined time period, the loggers continuously sample the accelerometers at a rate of 512Hz to ensure that no event goes uncaptured. At the end of the user defined time period, the largest acceleration value measured during the time period is recorded to the non-volatile memory (349,525 readings per axis).
The UltraShock is designed to be the complete solution for environmental monitoring. The device measures and records temperature, humidity, pressure and the peak acceleration levels for three axis (X, Y and Z) at the user selected readin