MadgeTech TCTemp2000 Thermocouple Based Temperature Recorder with LCD Display
Representative image only.
MadgeTech Mfr # TCTEMP2000 – Item # EW-23000-02
TCTemp2000 and RTDTemp2000
  • Large backlit LCD
  • Front panel keypad
  • Real-time operation
  • On-screen statistics
  • High-speed download
  • Wall mountable
  • 1 year warranty
$628.00USD / Each
sign in for your price.
$217.50 - $290.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Number of Channels1
  • Input TypeTemperature (thermocouple)
  • Temperature Accuracy (° F)±0.5
  • Temperature Resolution (° C)0.1
  • Temperature Accuracy (° C)±0.1 (0 to +50)
  • Memory Size131,071/Channel
  • Battery Life1 year
  • Display TypeLCD
  • Length (in)4 51/64
  • Height (in)3 19/64
  • Width (in)1 1/4
  • DescriptionThermocouple Based Temperature Recorder with LCD Display
  • Warranty1 year
1 Year Warranty
Key Features
  • Large backlit LCD
  • Front panel keypad
  • Real-time operation
  • On-screen statistics
  • High-speed download
  • Wall mountable
  • 1 year warranty
More About this Item
The TCTemp2000 is a thermocouple temperature logger that has an internal temperature sensor and one external thermocouple input. Compatible with J, K, T, E, R, S, B and N type thermocouples, the device can take 131,107 readings per channel.
The convenient LCD display allows for instant viewing of the current ambient reading and the current thermocouple reading. This provides users with a quick reference should the temperature need to be written down or used for another purpose. When using an infrared thermocouple, users can find the difference between the air temperature and surface temperature.
The RTDTemp2000 is a RTD based temperature logger, with LCD display. Its precise measurement readings and high accuracy make this logger useful in a variety of temperature monitoring applications such as calibration chamber monitoring, R&D, HVAC, and environmental studies.