MadgeTech TransiTempII Low Cost Temperature Data Logger
Representative image only.
MadgeTech Mfr # TRANSITEMPII – Item # EW-23000-18
TransiTempII, TransiTempII-RH, and Cryo-Temp
  • Programmable alarms and warnings
  • Instantaneous out of range indication
  • IP65 splash proof rated
  • Delayed Start
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Number of Channels1
  • Input TypeTemperature
  • Min Temperature (° C)-40
  • Max Temperature (° C)80
  • Temperature Accuracy (° F)±1.0
  • Temperature Resolution (° C)0.1
  • Temperature Accuracy (° C)±0.5
  • Memory Size32,767
  • Battery Life1 year - factory replacement
  • Length (in)3 19/64
  • Height (in)2 7/64
  • Width (in)13/32
  • DescriptionLow Cost Temperature Recorder
  • Warranty1 year
1 Year WarrantyConformite EuropeenneIP 65
Includes
Lanyard.
Key Features
  • Programmable alarms and warnings
  • Instantaneous out of range indication
  • IP65 splash proof rated
  • Delayed Start
  • 1 year warranty
More About this Item
These splash proof temperature data loggers featuring a highly precise semiconductor temperature sensor and an ABS plastic enclosure. This affordable, N.I.S.T. traceable device is ideal for monitoring and recording the temperature of all types of perishable products, including food items and pharmaceutical goods, both in storage and in transit.
The three available devices are equipped with three status LED indicators to alert you to the temperature status. Green signals safe conditions, yellow indicates warning limits have been exceeded, and red denotes alarm limits have been exceeded. The units can be programmed through software to start immediately, delayed start for up to 30 days, or be set to start using the magnetic switch.
The TransiTempII uses the IFC300 USB docking station for quick data download (model 23000-34 sold separately).