Manual Pinch Valve, 5/32" to 1/4" tube OD
Representative image only.
Item # EW-98002-00
$55.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Pinch Valve Tube Size5/32" to 1/4" OD
  • Connection TypePinch Valve for Tubing
  • DescriptionManual Pinch Valve, 5/32" to 1/4" tube OD
More About this Item
Fluid contacts only the tubing—ideal for use in pharmaceutical and food industries where eliminating contamination is critical. Obtain precise and repeatable flow rates using easy-to-read metered scale on valve body. Valves are made of Delrin®. Tubing is not included—order flexible tubing with durometer (Shore A) of 60 or less from our "Tubing" section.
Call Us At1-847-549-7600
Live chat