Mark-10 AC1095-1 Double Column Extension; 6"
Representative image only.
Mark-10 Mfr # ESM303-002-1 – Item # EW-25303-13
Configurable for your application needs!
  • Requires Mark-10 Motorized Test Stand Before Purchase
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionDouble Column Extension; 6\"
  • Warranty3 year
3 Year WarrantyConformite Europeenne
Key Features
  • Requires Mark-10 Motorized Test Stand Before Purchase
  • 3 year warranty
More About this Item
Accommodate up to a 20 in. [508 mm] wide sample, or ø21.5 in [ø546 mm] round sample.