Mark-10 G1053 Bottle cap grip, fits diameters of 0.65 to 1.8"
Representative image only.
Mark-10 Mfr # G1053 – Item # EW-59847-15
Safely secure objects during torque testing
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionBottle cap grip, fits diameters of 0.65 to 1.8\"
  • Warranty3 year
3 Year Warranty
Key Features
  • 3 year warranty
More About this Item
This grip effectively secures round objects during torque and force testing. Reversible serrated jaws accommodate samples with diameters of 0.65 to 1.55 in [16.5 to 39.4 mm] while an ergonomic, rubberized knob easily engages and disengages samples. A precision Acme screw provides durability and longevity over repeated engagements.
This grip can be used either manually with an R50 torque sensor or TT03 torque gauge, or in test stand controlled applications with the TSTM-DC torque test stand.
A hex tail with female threads permits mounting to a torque gauge, force gauge, or test stand.