Mark-10 MR51-20Z Torque Sensor, 20 ozFin / 1400 gFcm / 14 kgFmm / 140 Nmm / 14 Ncm
Representative image only.
Mark-10 Mfr # MR51-20Z – Item # EW-25302-31
Outstanding overload protection
  • Interchangeable chucks (order separately)
  • Warranty: 3 years
  • Outstanding overload protection
  • Interchangeable chucks (order separately)
  • Warranty: 3 years
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (oz-in)20
  • Capacity (g-cm)1400
  • Resolution (oz-in)0.01
  • Resolution (g-cm)1
  • Resolution (N-cm)0.01
  • Accuracy±0.20% of full scale + indicator
  • DescriptionPlug & Test Universal Torque Sensor, Interchange Grips; 14 N cm
3 Year WarrantyConformite Europeenne
Key Features
  • Interchangeable chucks (order separately)
  • Warranty: 3 years
  • Outstanding overload protection
  • Interchangeable chucks (order separately)
  • Warranty: 3 years
More About this Item
A great solution for general bi-directional torque testing applications requiring different sample sizes. Three interchangeable chuck capacities and bit holder are available separately.
A great solution for general bi-directional torque testing applications requiring different sample sizes. Three interchangeable chuck capacities and bit holder are available separately.