Mark-10 R54 Plug & Test Wrench Extension Torque Sensor; 110 N m
Representative image only.
Mark-10 Mfr # MR55-1000 – Item # EW-25302-50
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (lb-in)1000
  • Capacity (N-cm)110
  • Capacity (lb-ft)80
  • Resolution (lb-in)1
  • Resolution (lb-ft)0.1
  • Resolution (N-m)0.1
  • Accuracy±0.6% of full scale + indicator
  • DescriptionPlug & Test Wrench Extension Torque Sensor; 110 N m
  • Warranty3 year
3 Year WarrantyConformite Europeenne
Key Features
  • 3 year warranty
More About this Item
Convert any wrench to a torque tool with this sensor, or use it as a component of a torque testing system.
Convert any wrench to a torque tool with this sensor, or use it as a component of a torque testing system.