Mark-10 R54 Plug & Test Wrench Extension Torque Sensor; 550 N m
Representative image only.
Mark-10 Mfr # MR55-5000 – Item # EW-25302-51
$1,195.00USD / Each
For better pricing please sign in.
$65.00 - $246.00USD / Each
for an additional
$65.00USD / Each
for an additional
$246.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (lb-in)5000
  • Capacity (N-cm)550
  • Capacity (lb-ft)400
  • Resolution (lb-in)5
  • Resolution (lb-ft)0.5
  • Resolution (N-m)0.5
  • Accuracy±0.6% of full scale indicator
  • DescriptionPlug & Test Wrench Extension Torque Sensor; 550 N m
More About this Item
Convert any wrench to a torque tool with this sensor, or use it as a component of a torque testing system.
Convert any wrench to a torque tool with this sensor, or use it as a component of a torque testing system.