Marsh Bellofram 2KSNNF42DF31500 Pressure Trandsucer, I to P;3-15 psi Output, 4-20 mA Input
Representative image only.
Marsh Bellofram Mfr # 2KSNNF42DF31500 – Item # EW-68826-24
±0.25% full scale accuracy
  • 4 to 20 mA Input
  • High and low split ranging
  • Field-selectable features
  • Direct and reverse acting input and output
  • 18 month warranty
PRODUCT OPTIONS
$815.00USD / Each
$152.00 - $225.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Min Exhaust Flow Rate (SCFM)1
  • Forward Flow Capacity (SCFM)24
  • Forward Flow Capacity (LPM)677
  • Min Operating Temperature (° F)-20
  • Max Operating Temperature (° F)160
  • Min Operating Temperature (° C)-28
  • Max Operating Temperature (° C)71
  • Control Range (low) (PSI)3
  • Control Range (high) (PSI)15
  • Process Connection NPT(F) (in)1/4
  • Electrical Connections½\" conduit
  • DescriptionPressure Trandsucer, I to P;3-15 psi Output, 4-20 mA Input
  • Warranty18 months
18 Month WarrantyCanadian Standards AssociationFM ApprovedIP 65ISO9001:2000 Certified Supplier
Key Features
  • 4 to 20 mA Input
  • High and low split ranging
  • Field-selectable features
  • Direct and reverse acting input and output
  • 18 month warranty
More About this Item
Transducers are designed for valve actuating, air cylinders, and controllers. The 4 to 20 mA current input is conditioned to provide a signal directly proportional to the desired pressure output. The piezo-ceramic actuator serves as a control link between electrical input and pressure output. As voltage increases, the force applied by the actuator increases, so as to restrict nozzle bleed and thus increase pilot pressure. The increased pilot pressure applied to the diaphragm causes opening of the supply valve increasing output pressure.