Martel 1010056 Accessory Kit for MECP10K includes carrying case, test hose and fittings
Representative image only.
Martel Mfr # 1010056 – Item # EW-68580-08
Capable of generating both vacuum and pressure
  • Generate –27 ”Hg to 100 or 600 PSI, model dependent
  • Generate hydraulic pressure from 0 to 10,000 psi
  • Completely isolated vernier prevents heat transfer from hand
  • Precision valve for bleed control
  • Ergonomic design makes it easy to pump, even at high pressure
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionAccessory Kit for MECP10K includes carrying case, test hose and fittings
  • Warranty1 Year
1 Year Warranty
Key Features
  • Generate –27 ”Hg to 100 or 600 PSI, model dependent
  • Generate hydraulic pressure from 0 to 10,000 psi
  • Completely isolated vernier prevents heat transfer from hand
  • Precision valve for bleed control
  • Ergonomic design makes it easy to pump, even at high pressure
  • 1 year warranty
More About this Item
These high-performance, hand-operated pumps generate both pressure and vacuum for precise testing of pressure instruments. The output pressure can be fine tuned using the vernier control. A precision needle valve allows controlled venting for exacting pressure calibration requirements. All pumps are offered in a convenient kit with hard-sided carrying case and flexible connection hose with appropriate fittings.
MECP100 and MECP500 pneumatic pumps allow pressures in excess of 100 or 500 psi to be generated, respectively, with just a few strokes of the pump. By changing the switching valve to vacuum, a range of -27" Hg is available. They also feature user-friendly, field serviceable, and replaceable check valves.
MECP10K Hydraulic Pressure Test Pump works with a variety of hydraulic fluids or even water to easily generate test pressures up to 10,000 psi. It uses a unique fully adjustable stroke control to allow for fast priming or filling of test systems, and then switching as needed to a smaller stroke for easier pumping at high pressures.