Marvel Scientific MS24RASFRW 24" Flammable Material Refrigerator, 6.1 cu ft, 120V

Representative image only.
Marvel Scientific Mfr # MS24RASFRW – Item # EW-44766-07
Engineered and built to meet or exceed the stringent standards required by science and industry
  • Choose from general-purpose, flammable, and hazardous storage units

PRODUCT OPTIONS

$4,750.00USD / Each
Stay Connected - Monitor Your Refrigerator or Freezer Temperatures

SPECS & DESCRIPTION

Specifications & Description

  • Refrigerator Capacity (cu ft)6.1
  • Refrigerator Capacity (Liters)173
  • Min Temperature - Refrigerator (° F)33
  • Max Temperature - Refrigerator (° F)40
  • Min Temperature - Refrigerator (° C)1
  • Max Temperature - Refrigerator (° C)5
  • Temperature ControlMechanical Thermostat
  • Refrigerator Interior Width (in)21
  • Refrigerator Interior Height (in)28
  • Refrigerator Interior Depth (in)20
  • Refrigerator Interior Width (cm)53
  • Refrigerator Interior Height (cm)71
  • Refrigerator Interior Depth (cm)53
  • Exterior Width (in)23
  • Exterior Height (in)34
  • Exterior Depth (in)26
  • Exterior Width (cm)61
  • Exterior Height (cm)86
  • Exterior Depth (cm)66
  • Number of Doors1
  • Number of Shelves2, 3 door shelves
  • Power (VAC)115
  • Power (Hz)60
  • Power (amps)2.5
  • Description24" Flammable Material Refrigerator, 6.1 cu ft

Includes

adjustable and removable wire shelves. Commercial power cord and plug, except for hazardous location (explosion-proof) models which require hard-wiring.

Key Features

  • Choose from general-purpose, flammable, and hazardous storage units

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat