Masterflex Barcode Scanner
Representative image only.
Masterflex Item # EW-24507-18
Build a complete single-use flow sensor system customizable to your application
  • Modular interchangeable flow sensor systems with single-use design
  • Maintain accuracy and repeatability by not having to manually enter K-factor—saving time and eliminating errors
  • Connect to a controller, display, or computer via 5 to 30 V square wave pulse output for easy measurement readings
$748.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Min Operating Temperature (° F)14
  • Min Operating Temperature (° C)-10
  • Max Operating Temperature (° F)140
  • Max Operating Temperature (° C)60
  • Power5 to 24 VDC
  • Output (mA)No
  • Output (Voltage)Yes
  • OutputOne 5 to 30 volt square wave pulse
  • Input SignalOne 5 to 30 volt square wave pulse
  • Display TypeNone
  • DescriptionBarcode Scanner
Key Features
  • Modular interchangeable flow sensor systems with single-use design
  • Maintain accuracy and repeatability by not having to manually enter K-factor—saving time and eliminating errors
  • Connect to a controller, display, or computer via 5 to 30 V square wave pulse output for easy measurement readings
More About this Item
The Masterflex® single-use flow sensor system is comprised of four components. Select the flow sensor, flow tubes, barcode scanner, and output that suit your application needs for a customizable approach to single-use flow measurement.
Each flow tube has an attached label that indicates the lot number, serial number, K-factor, and 2-D bar code with all the calibration data. The barcode can quickly be read by the barcode scanner making the calibration process unnecessary when exchanging a flow tube. Eliminate long downtimes and extra calibration while ensuring an accurate measurement with the barcode scanner.
Flow tube sensor, flow tubes, output options, and calibration certificates sold separately.