Masterflex DB15 Male Connector; for Creating Your Own Cable
Representative image only.
Masterflex Item # EW-07595-52
Create your own cable for optimization of your Masterflex® drive
  • Choose the correct connection for your Masterflex® pump
PRODUCT OPTIONS
$74.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionDB15 male connector; for creating your own cable.
Key Features
  • Choose the correct connection for your Masterflex® pump
More About this Item
Combine these connectors with 20-22 gauge shielded wire to create your own interface cable. Maximum cable length is 1000 ft (300 m).