Masterflex Drum Pump Replacement Motor, TEFC; 110V/60Hz

Representative image only.
Masterflex Item # EW-70604-13
Safety transfer a variety of liquids from drums, tanks, and small storage vessels
  • Use with Masterflex® Complete Drum and Container Pumps
  • Powered by 110V/60Hz TEFC motor

PRODUCT OPTIONS

$1,057.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Motor TypeTotally Enclosed, Fan Cooled (TEFC)
  • Power (VAC)110
  • Power (Hz)60
  • DescriptionReplacement motor, TEFC, 110 VAC, 60 Hz

Key Features

  • Use with Masterflex® Complete Drum and Container Pumps
  • Powered by 110V/60Hz TEFC motor

More About this Item

Drum and container pumps are engineered to safely transfer a variety of liquids from drums, tanks and small storage vessels. These Masterflex® drum pump replacement motors are one component of a system that includes a pump tube and motor (order pump tubes separately). Motors are equipped with variable-speed control for added safety and control.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat