Masterflex Electro-Optic Point-Level Sensor, Nickel-Plated Steel/Glass, Dry Sink
Representative image only.
Masterflex Item # EW-32772-32
No clog or wear—contain no moving parts
  • Detect liquids regardless of conductivity, color, or specific gravity
  • Economical and compact — built-in solid-state switching electronics
  • One-unit installation — simply screws into the top or side of your tank
PRODUCT OPTIONS
$258.60USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Relay Output30 VDC at 40 mA max
  • Wetted MaterialNickel-plated Carbon steel/Fused glass
  • Process Mount1/2"NPT(M)
  • Max Process Temperature (° C)100
  • Max Pressure (PSI)2500
  • Power (VDC)12
  • DescriptionElectro-Optic Point-Level Sensor, Nickel-Plated Steel/Glass, Dry Sink; 1/2"NPT(M)
Key Features
  • Detect liquids regardless of conductivity, color, or specific gravity
  • Economical and compact — built-in solid-state switching electronics
  • One-unit installation — simply screws into the top or side of your tank
More About this Item
These dependable, electro-optic level sensors contain no moving parts to clog or wear. Sensors offer ±1 mm repeatability and are available in a wide variety of materials to meet even the most demanding applications.
Installation is simple and quick through the tank top, bottom, or side. The switches can be used for low-, intermediate-, or high-liquid level detection. Solid state switching ensures dependability over a long service life. Order controller separately.