Masterflex Extension Cable for L/S® and I/P® Digital Modular Drives; 9 ft
Representative image only.
Masterflex Item # EW-77301-82
Increase modular drive flexibility with extended cable lengths
  • Easily connect to your Masterflex® pump with connectors on both ends
PRODUCT OPTIONS
$525.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionExtension Cable for L/S and I/P Digital Modular Drives; 9 ft
Key Features
  • Easily connect to your Masterflex® pump with connectors on both ends
More About this Item
Masterflex extension cables can be used with L/S and I/P modular drives to extend the distance between drive motor and controller by 9 ft (2.7 m) to increase installation flexibility. Do not extend more than 25ft.
This extension cable is compatible with L/S® pump models 77301-40, -50, -60, and -70; and with I/P® pump models 07594-00 and -10.