Masterflex Fitting, Nylon, Straight, Hose Barb Reducer, 1/2" ID x 3/8" ID; 100/PK
Representative image only.
Masterflex Item # EW-30626-43
Best for low-pressure applications
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
PRODUCT OPTIONS
$83.50USD / Pkg of 100
BUY MORE & SAVE
  • 1-9$83.50USD / Pkg of 100
  • 10-49$75.15USD / Pkg of 100
  • 50-99$70.98USD / Pkg of 100
  • 100+$66.80USD / Pkg of 100
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialNylon
  • Sterilization MethodEthylene oxide (ETO) only
  • Connection TypeBarb
  • Barb Size / Tubing Id (Port 1) (in)1/2
  • Barb Size / Tubing Id (Port 2) (in)3/8
  • DescriptionReducing Connector, Nylon, 1/2" x 3/8", 100 PACK
Key Features
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Temperture range -50 to 250°F (-46 to 121°C)
  • Connection Type: 1/2" ID x 3/8" ID
More About this Item
These economical straight barbed reducing fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.