Masterflex Fitting, Nylon, Straight, Hose Barb to Threaded Adapter, 3/8" ID x 1/4" NPT(F); 10/PK
Representative image only.
Masterflex Item # EW-31702-22
No tools needed for assembly
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
PRODUCT OPTIONS
$19.30USD / Pkg of 10
BUY MORE & SAVE
  • 1-9$19.30USD / Pkg of 10
  • 10-49$17.37USD / Pkg of 10
  • 50-99$16.41USD / Pkg of 10
  • 100+$15.44USD / Pkg of 10
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialNylon
  • Sterilization MethodEthylene oxide (ETO) only
  • Connection TypeBarb x NPT(F)
  • Thread size NPT(F) (port 2) (in)1/4
  • Barb Size / Tubing Id (Port 1) (in)3/8
  • DescriptionThreaded adapter Female 1/4 x 3/8" pack of 10
Key Features
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Connection Type: 3/8" ID x 1/4" NPT(F)
More About this Item
Barbed fittings are designed for quick and easy assembly without tools. Combine these simple and economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).