Masterflex Fitting, Nylon, Straight, Hose Barb Union, 3/4" ID; 10/PK
Representative image only.
Masterflex Item # EW-30621-82
Quick and easy assembly without tools
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
PRODUCT OPTIONS
$26.50USD / Pkg of 10
BUY MORE & SAVE
  • 1-9$26.50USD / Pkg of 10
  • 10-49$23.85USD / Pkg of 10
  • 50-99$22.53USD / Pkg of 10
  • 100+$21.20USD / Pkg of 10
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialNylon
  • Sterilization MethodEthylene oxide (ETO)
  • Connection TypeBarb
  • Barb Size / Tubing Id (Port 1) (in)3/4
  • Barb Size / Tubing Id (Port 2) (in)3/4
  • DescriptionFitting, Nylon, Straight, Hose Barb Union, 3/4" ID; 10/PK
Key Features
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Chemical resistant
  • Connection Type: 3/4" ID
More About this Item
These economical straight barbed fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.