Masterflex Fitting, Nylon, Straight, Snap Luer Lock Ring; 25/PK
Representative image only.
Masterflex Item # EW-45505-74
Easily transition to a luer fitting
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
Free Certificate of Comliance
$11.30USD / Pkg of 25
BUY MORE & SAVE
  • 1-9$11.30USD / Pkg of 25
  • 10-49$10.17USD / Pkg of 25
  • 50-99$9.61USD / Pkg of 25
  • 100+$9.04USD / Pkg of 25
SPECS & DESCRIPTION
Specifications & Description
  • MaterialNylon
  • DescriptionSnap Luer Lock Ring for Male Slip Luers, Nylon, 25/pk
Certificate of ComplianceCole-Parmer Exclusive
Key Features
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Snap luer lock ring for male slip luers
More About this Item
Easily complete your luer fittings systems with these premium connectors. Included are some special configurations available only in polycarbonate. The snap luer lock ring is for use with male luer by wide-bore hose barb fittings. Nylon material has good chemical resistance and can be sterilized via autoclave, ethylene oxide, or gamma irradiation.