Masterflex Fitting, Polycarbonate, One-Way Stopcock, Male Luer Lock, Non-Sterile; 100/PK
Representative image only.
Masterflex Item # EW-30600-05
Regulate flow without any leaking
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
PRODUCT OPTIONS
$232.00USD / Pkg of 100
BUY MORE & SAVE
  • 1-9$232.00USD / Pkg of 100
  • 10-49$208.80USD / Pkg of 100
  • 50-99$197.20USD / Pkg of 100
  • 100+$185.60USD / Pkg of 100
SPECS & DESCRIPTION
Specifications & Description
  • Outlet PortMale Luer Lock
  • Inlet PortFemale Luer
  • Wetted MaterialsPolycarbonate
  • Flow PatternOne-Way
  • DescriptionStopcocks with Luer connection, male lock, 1-way; 100/pack, Non-sterile
Key Features
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • One-way stopcock/flow diverter with male luer lock connection
  • HDPE handle rotates 90°
More About this Item
Choose stopcocks with either a male luer lock or a male luer slip. Handle rotates 90° on one-way stopcocks, 180° on three-way stopcocks, and 360° on four-way stopcocks. All have PC body and HDPE stem.