Masterflex Fitting, Polycarbonate, Straight, Female Luer to Smooth Bore Barb Hose Adapter, 3/32" ID; 25/PK
Representative image only.
Masterflex Item # EW-45501-52
Ensure your process integrity with full material traceability
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
PRODUCT OPTIONS
Free Certificate of Comliance
$15.90USD / Pkg of 25
BUY MORE & SAVE
  • 1-9$15.90USD / Pkg of 25
  • 10-49$14.31USD / Pkg of 25
  • 50-99$13.52USD / Pkg of 25
  • 100+$12.72USD / Pkg of 25
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolycarbonate
  • Number of Ports2
  • Connection Type-Port 1Female luer
  • Barb Size-Port 2 (in)15/16
  • DescriptionFemale luer x 3/32" hose barb, smooth bore, PC, 25/pk
Certificate of ComplianceCole-Parmer Exclusive
Key Features
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Designed for quick and easy assembly with flexible tubing
  • Connection Type: Female Luer to smooth Bore Barb Hose Adapter, 3/32" ID
More About this Item
These luer fittings are ideal for a variety of applications, including syringes or small tubing. Barbed adapters feature three styles of barbs: standard, wide-bore, and low-profile. Wide-bore barbs offer enhanced fluid flow through the fitting and are usable with any flexible tubing. Low-profile barbs are ideal for use with semi-rigid tubing, such as nylon, polyethylene, or polypropylene. Manufactured in compliance with ISO 594-1 standards, these fittings are cleaned, inspected and fully traceable. All parts meet tight dimensional tolerances for increased reliability and process purity.
Hose barbs are available from 1/16" to 1/4" with either male or female luer connections. Male luer connections are available with either integral lock ring or separate lock ring.