Masterflex Fitting, Polycarbonate, Straight, Male Slip Luer to Threaded Adapter, 1/4-28 UNF; 10/PK
Representative image only.
Masterflex Item # EW-30505-55
Easily transition between luer fittings and UNF or NPT threads
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
PRODUCT OPTIONS
$16.00USD / Pkg of 10
BUY MORE & SAVE
  • 1-9$16.00USD / Pkg of 10
  • 10-49$14.40USD / Pkg of 10
  • 50-99$13.60USD / Pkg of 10
  • 100+$12.80USD / Pkg of 10
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolycarbonate
  • Number of Ports2
  • Connection Type-Port 1Male luer slip
  • Connection Type-Port 21/4-28 UNF
  • DescriptionMale slip luer x 1/4-28 UNF, Polycarbonate, 10 Pack
Key Features
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Connection Type: 1/4-28 UNF
More About this Item
These adapters transition between luer fittings and UNF or NPT threads. Both male and female luer connections are available. All threaded connections are male thread.