Masterflex Fitting, Polycarbonate, Three-Way, Large-Bore Stopcocks with Male Luer Lock, Non-Sterile; 100/PK
Representative image only.
Masterflex Item # EW-30600-25
Enhance fluid flow with a larger bore than standard luer stopcocks
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
PRODUCT OPTIONS
$234.00USD / Pkg of 100
BUY MORE & SAVE
  • 1-9$234.00USD / Pkg of 100
  • 10-49$210.60USD / Pkg of 100
  • 50-99$198.90USD / Pkg of 100
  • 100+$187.20USD / Pkg of 100
SPECS & DESCRIPTION
Specifications & Description
  • Outlet PortMale Luer Lock
  • Inlet PortFemale Luer
  • Wetted MaterialsPolycarbonate
  • Flow PatternThree-Way
  • Description3-way male lock stopcock, 100/pack, Non-sterile
Key Features
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Three-way flow pattern
More About this Item
Large-bore adapters are perfect for handling higher flow volumes in process while using connections similar to luer systems.