Masterflex Fitting, Polycarbonate, Two-Way, High-Pressure Stopcocks with Male Luer Lock to Female Luer, Non-Sterile; 10/PK
Representative image only.
Masterflex Item # EW-30526-32
Pressure rating up to 1050 psi
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
PRODUCT OPTIONS
$44.75USD / Pkg of 10
BUY MORE & SAVE
  • 1-9$44.75USD / Pkg of 10
  • 10-49$40.28USD / Pkg of 10
  • 50-99$38.04USD / Pkg of 10
  • 100+$35.80USD / Pkg of 10
SPECS & DESCRIPTION
Specifications & Description
  • Outlet PortMale Luer Lock
  • Inlet PortFemale Luer
  • Max Pressure (PSI)1050
  • Wetted MaterialsPolycarbonate
  • Flow PatternTwo-Way
  • DescriptionHigh Pressure Stopcock, 1050 psi max, 2-way, 10 pack, Non-sterile
Key Features
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Connection Type: two-way flow pattern
More About this Item
Stopcock fittings feature luer connections.