Masterflex Fitting, Polyethylene, Straight, Male Threaded Adapter, 1/8" NPT(M); 100/PK
Representative image only.
Masterflex Item # EW-30627-24
Maintain a permanent connection
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
PRODUCT OPTIONS
Free Certificate of Comliance
$93.50USD / Pkg of 100
BUY MORE & SAVE
  • 1-9$93.50USD / Pkg of 100
  • 10-49$84.15USD / Pkg of 100
  • 50-99$79.48USD / Pkg of 100
  • 100+$74.80USD / Pkg of 100
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialHDPE
  • Sterilization MethodEthylene oxide (ETO) only
  • Connection TypeNPT(M)
  • Thread size NPT(M) (port 1) (in)1/8
  • Thread size NPT(M) (port 2) (in)1/8
  • Barb Size / Tubing Id (Port 1) (in)1/8
  • DescriptionFitting, Nipple, Polyethylene, 1/8" x, 100 PACK
Certificate of Compliance
Key Features
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Connection Type: 1/8" NPT(M)
  • Constructed of durable polyethylene
More About this Item
Threaded fittings provide a more permanent fluid path. Choose the size and material to meet your application needs.