Masterflex Fitting, Polypropylene, RQX Male Connector Plug; 1/Ea
Representative image only.
Masterflex Item # EW-40618-56
Disconnect and reconnect your flow path with ease
  • Large ergonomic button and thumb pad with secure latch and optional integrated locking mechanism
  • Several connector configurations available in commonly used polypropylene and polycarbonate materials
  • Plugs and caps available to maintain fluid path integrity
PRODUCT OPTIONS
$12.09USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPolypropylene
  • Connection TypeQuick-Disconnect x Plug
  • DescriptionFitting, Polypropylene, RQX Male Connector Plug; 1/Ea
Key Features
  • Large ergonomic button and thumb pad with secure latch and optional integrated locking mechanism
  • Several connector configurations available in commonly used polypropylene and polycarbonate materials
  • Plugs and caps available to maintain fluid path integrity
More About this Item
RQ Series offers a cost effective solution for flexible tubing connections in single use systems. The connector set features a user-friendly interface that includes a large guarded button for a secure lock to guard against disconnects. Use these fittings to quickly connect and disconnect from your flow line. Simply order a body (female half) and an insert (male half).