Masterflex Fitting, Polypropylene, Straight, Male Luer to Hose Barb Adapter, 1/4" ID; 25/PK
Representative image only.
Masterflex Item # EW-30800-26
Easily transition to a luer fitting
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
PRODUCT OPTIONS
$20.75USD / Pkg of 25
BUY MORE & SAVE
  • 1-9$20.75USD / Pkg of 25
  • 10-49$18.68USD / Pkg of 25
  • 50-99$17.64USD / Pkg of 25
  • 100+$16.60USD / Pkg of 25
Often Bought with…
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolypropylene
  • Number of Ports2
  • Connection Type-Port 1Male luer slip
  • Connection Type-Port 2Hose barb
  • Barb Size-Port 2 (in)1/4
  • DescriptionADCF Male Luer to 1/4" L Barb Adapter, PP, 25/pk
Certificate of Compliance
Key Features
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Connection Type: Male luer to 1/4" hose barb with 1.103" overall length
More About this Item
Manufactured of animal-free natural polypropylene, these luers meet Class VI specifications. Luers offer superior chemical resistance and are suited for ETO and gamma sterilization.