Masterflex Fitting, Polypropylene, Straight, Male Luer to Hose Barb Adapter, 3/32" ID; 25/PK
Representative image only.
Masterflex Item # EW-30800-28
Easily transition to a luer fitting
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
PRODUCT OPTIONS
$19.80USD / Pkg of 25
BUY MORE & SAVE
  • 1-9$19.80USD / Pkg of 25
  • 10-49$17.82USD / Pkg of 25
  • 50-99$16.83USD / Pkg of 25
  • 100+$15.84USD / Pkg of 25
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolypropylene
  • Number of Ports2
  • Connection Type-Port 1Male luer lock
  • Connection Type-Port 2Hose barb
  • Barb Size-Port 2 (in)3/32
  • DescriptionAnimal Free Male Luer Adapter, 3/32", PP, 25/pk
Certificate of Compliance
Key Features
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Connection Type: Male luer to 3/32" hose barb with 0.068" diameter orifice
More About this Item
Manufactured of animal-free natural polypropylene, these luers meet Class VI specifications. Luers offer superior chemical resistance and are suited for ETO and gamma sterilization.