Masterflex Fitting, Polypropylene, Straight, Sanitary Clamp to Hose Barb Adapter, 1 1/2" Tri-Clamp® x 1/2" ID; 6/PK
Representative image only.
Masterflex Item # EW-40618-37
Maintain sanitary conditions
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
PRODUCT OPTIONS
$99.00USD / Pkg of 6
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPolypropylene
  • Barb Size / Tubing Id (Port 2) (in)1/2
  • Sanitary Clamp Size (Port 1) (in)1.5
  • DescriptionFitting, Polypropylene, Straight, Sanitary Clamp to Hose Barb Adapter, 1 1/2" Tri-Clamp® x 1/2" ID; 6/PK
Key Features
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Meet requirements for USP Class VI, FDA, REACH, and California Proposition 65
  • Surface finishes to 15 micron minimizes adhesion or entrapment
  • Packaged to exacting tolerances under ISO Class 7 cleanroom conditions
  • Compatible with other Tri-Clamp® connections
More About this Item
The sanitary Tri-Clamp barbed adapters are ideal for use in the biopharmaceutical, food and beverage, and single-use disposable industries. The fittings are made from animal-derived ingredient-free and phthalate-free polypropylene (PP) and PVDF. They have excellent chemical resistance and may be sterilized by gamma irradiation, EtO, E-beam, or autoclave. The PVDF fittings withstand higher temperatures than the PP fittings and offer extremely low surface tension for lower protein and lipid binding.