Masterflex Fitting, Polypropylene, Tee, Female Luer to Female Luer to Female Luer Adapter; 25/PK
Representative image only.
Masterflex Item # EW-45508-56
Full material traceability to ensure process integrity
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
PRODUCT OPTIONS
Free Certificate of Comliance
$32.00USD / Pkg of 25
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolypropylene
  • Number of Ports3
  • Connection Type-Port 1Female luer
  • Connection Type-Port 2Female luer
  • Connection Type-Port 3Female luer
  • DescriptionLuer Adapter Tees, Female x Female x Female, PP, 25/Pk
Certificate of Compliance
Key Features
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Manufactured from 100% virgin raw materials in a Class 100,000 environment
  • Connection Type: Female Luer to Female Luer to Female Luer
More About this Item
These luer fittings are ideal for a variety of applications, including syringes or small tubing. Manufactured in compliance with ISO 594-1 standards, these fittings are cleaned, inspected and fully traceable. All parts meet tight dimensional tolerances for increased reliability and process purity.