Masterflex Fitting, PVDF, Straight, Female Luer to Cap Adapter, 25/PK
Representative image only.
Masterflex Item # EW-45512-28
Full material traceability to ensure process integrity
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
PRODUCT OPTIONS
$31.50USD / Pkg of 25
BUY MORE & SAVE
  • 1-9$31.50USD / Pkg of 25
  • 10-49$28.35USD / Pkg of 25
  • 50-99$26.78USD / Pkg of 25
  • 100+$25.20USD / Pkg of 25
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPVDF
  • Number of Ports1
  • Connection Type-Port 1Female luer
  • Connection Type-Port 2Cap
  • DescriptionLuer Accessory, Female Luer Cap, PVDF, 25/Pk
Certificate of Compliance
Key Features
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Manufactured from 100% virgin raw materials in a Class 100,000 environment
  • Connection Type: Female Luer x Cap
More About this Item
These luer fittings are ideal for a variety of applications, including syringes or small tubing. Manufactured in compliance with ISO 594-1 standards, these fittings are cleaned, inspected and fully traceable. All parts meet tight dimensional tolerances for increased reliability and process purity.