Masterflex Fitting, PVDF, Straight, Male Luer to Threaded Adapter, 1/8-27 NPT; 25/PK
Representative image only.
Masterflex Item # EW-45513-86
Easily transition between luer fittings and UNF or NPT threads
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
PRODUCT OPTIONS
$66.50USD / Pkg of 25
BUY MORE & SAVE
  • 1-9$66.50USD / Pkg of 25
  • 10-49$59.85USD / Pkg of 25
  • 50-99$56.53USD / Pkg of 25
  • 100+$53.20USD / Pkg of 25
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPVDF
  • Number of Ports2
  • Connection Type-Port 1Male luer lock
  • Connection Type-Port 21/8-27" thread
  • DescriptionAdapter, PVDF, Male Luer To 1/8-27 Thread, 25/pack
Certificate of Compliance
Key Features
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Connection Type: Male Luer Lock x 1/8-27 " NPT
More About this Item
These adapters transition between luer fittings and UNF or NPT threads. Both male and female luer connections are available. All threaded connections are male thread.