Masterflex Fitting, PVDF, Straight, Male Threaded Adapter, 3/4" NPT(M); 10/PK
Representative image only.
Masterflex Item # EW-40615-27
Maintain a permanent connection
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
PRODUCT OPTIONS
$65.60USD / Pkg of 10
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPVDF
  • Sterilization MethodAutoclave or ethylene oxide (ETO)
  • Connection TypeNPT(M)
  • Thread size NPT(M) (port 1) (in)3/4
  • Thread size NPT(M) (port 2) (in)3/4
  • Flow PatternStraight
  • Fitting Length (in)1 7/8
  • Fitting Width (in)1 5/16
  • Fitting Width (mm)33
  • DescriptionFitting, PVDF, Straight, Male Threaded Adapter, 3/4" NPT(M); 10/PK
Key Features
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
More About this Item
Threaded fittings provide a more permanent fluid path. Choose the size and material to meet your application needs.