Masterflex Fitting, PVDF, Straight, Sanitary Clamp to Hose Barb Adapter, Mini Tri-Clamp® x 3/8" ID; 6/PK
Representative image only.
Masterflex Item # EW-31200-53
Maintain sanitary conditions
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
PRODUCT OPTIONS
Free Certificate of Comliance
$65.50USD / Pkg of 6
BUY MORE & SAVE
  • 1-3$65.50USD / Pkg of 6
  • 4-9$58.95USD / Pkg of 6
  • 10-19$55.68USD / Pkg of 6
  • 20+$52.40USD / Pkg of 6
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPVDF
  • Barb Size / Tubing Id (Port 2) (in)3/8
  • DescriptionBarb Sanitary Adapter, Mini Tri-Clamp x 3/8" Hose Barb, PVDF; 6/pk
Certificate of Compliance
Key Features
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Meet requirements for USP Class VI, FDA, REACH, and California Proposition 65
  • Surface finishes to 15 micron minimizes adhesion or entrapment
  • Packaged to exacting tolerances under ISO Class 7 cleanroom conditions
  • Compatible with other Tri-Clamp® connections
  • Connection Type: Mini Tri-Clamp x 3/8" ID
More About this Item
The sanitary Tri-Clamp barbed adapters are ideal for use in the biopharmaceutical, food and beverage, and single-use disposable industries. The fittings are made from animal-derived ingredient-free and phthalate-free polypropylene (PP) and PVDF. They have excellent chemical resistance and may be sterilized by gamma irradiation, EtO, E-beam, or autoclave. The PVDF fittings withstand higher temperatures than the PP fittings and offer extremely low surface tension for lower protein and lipid binding.