Masterflex Fitting, PVDF, Straight, Sanitary Clamp to Threaded Adapter, 3/4" Mini to 3/8" NPT(F)
Representative image only.
Masterflex Item # EW-40619-57
Maintain sanitary conditions
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
PRODUCT OPTIONS
$197.31USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPVDF
  • Connection TypeSanitary x NPT(F)
  • Thread size NPT(F) (port 2) (in)3/8
  • Sanitary Clamp Size (Port 1) (in)3/4
  • DescriptionFitting, PVDF, Straight, Sanitary Clamp to Threaded Adapter, 3/4" Mini to 3/8" NPT(F)
Key Features
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Meet USP Class VI for pharmaceutical use and FDA, USDA, and 3A standards for food contact
  • All fittings are compatible with Tri-Clamp® connections
  • Excellent chemical resistance; low leachable characteristics
More About this Item
These fittings are ideal for use with the Masterflex L/S tubing pumps.