Masterflex Fitting, PVDF, Straight, Threaded Reducer, 1/8" NPT(M) x 1/4" NPT(M); 10/PK
Representative image only.
Masterflex Item # EW-30623-33
Quick and easy assembly
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
PRODUCT OPTIONS
Free Certificate of Comliance
$46.25USD / Pkg of 10
BUY MORE & SAVE
  • 1-9$46.25USD / Pkg of 10
  • 10-49$41.63USD / Pkg of 10
  • 50-99$39.31USD / Pkg of 10
  • 100+$37.00USD / Pkg of 10
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPVDF
  • Sterilization MethodAutoclave or ethylene oxide (ETO)
  • Connection TypeNPT(M)
  • Thread size NPT(M) (port 1) (in)1/8
  • Thread size NPT(M) (port 2) (in)1/4
  • DescriptionFitting, PVDF, Straight, Threaded Reducer, 1/8" NPT(M) x 1/4" NPT(M); 10/PK
Certificate of Compliance
Key Features
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Connection Type: 1/8" NPT(M) x 1/4" NPT(M)
  • Constructed of durable PVDF
More About this Item
Threaded fittings provide a more permanent fluid path. Use these fittings to reduce or enlarge connection sizes. Choose the size and material to meet your application needs.