Masterflex Flow Indicator, Brass Housing, Orange Rotor, 3/4" NPT(F); 30 GPM
Representative image only.
Masterflex Item # EW-32779-08
Easily view flow through clear plastic lens
  • Indicators feature a bright orange rotor that spins to provide visual verification
PRODUCT OPTIONS
$288.00USD / Each
$332.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Min Flow Rate (GPM)5
  • Max Flow Rate (GPM)30
  • Material O-RingViton®
  • Material HousingBrass
  • Wetted MaterialsRotor pin: Ceramic; Lens: Polysulfone
  • Min Operating Temperature (° F)-20
  • Min Operating Temperature (° C)-29
  • Max Operating Temperature (° F)212
  • Max Operating Temperature (° C)100
  • Pipe Size3/4" NPT(F)
  • Display TypeHigh Visibility Orange, Molded Nylon Rotor
  • DescriptionFlow Indicator; Brass, 3/4" NPT(F), Orange Rotor
Key Features
  • Indicators feature a bright orange rotor that spins to provide visual verification
More About this Item
Flow indicators feature a paddle-wheel that spins at a rate proportional to the flow. Lens can be removed with a 1/4 turn for easy cleaning. Use with fluids that have a viscosity less than 34 centipoise (150 µm filter is r ecommended for protection).
Indicators/Sensors feature a Hall-effect sensor that produces a 4.5 to 24 VDC pulse output. Maximum current source output is 70 mA, and frequency output range is approximately 25 Hz at low flow to 225 Hz at high flow.
Indicators/Switches offer extra level of reliability and protection with a built-in switch. Each Single Pole Double Throw (SPDT) switch is rated for 1 amp, 24 VDC resistive. Repeatability is ±2% ; accuracy is ±5%. An LED lights when the set point is reached for easy field calibration.