Masterflex Flow Indicator/Switch, Polypropylene Housing, 1/2" NPT(F), 20 GPM; 24 VDC
Representative image only.
Masterflex Item # EW-32880-32
Easily view flow through clear plastic lens
  • Indicators feature a bright orange rotor that spins to provide visual verification
PRODUCT OPTIONS
$298.90USD / Each
$332.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Max Flow Rate (GPM)20
  • Max Operating Temperature (° F)180
  • Max Operating Temperature (° C)82
  • Pipe Size1/2" NPT(F)
  • DescriptionFlow Sensor/Switch; Polypropylene, 1/2" NPT(F), 24 VDC
Key Features
  • Indicators feature a bright orange rotor that spins to provide visual verification
More About this Item
Flow indicators feature a paddle-wheel that spins at a rate proportional to the flow. Lens can be removed with a 1/4 turn for easy cleaning. Use with fluids that have a viscosity less than 34 centipoise (150 µm filter is r ecommended for protection).
Indicators/Sensors feature a Hall-effect sensor that produces a 4.5 to 24 VDC pulse output. Maximum current source output is 70 mA, and frequency output range is approximately 25 Hz at low flow to 225 Hz at high flow.
Indicators/Switches offer extra level of reliability and protection with a built-in switch. Each Single Pole Double Throw (SPDT) switch is rated for 1 amp, 24 VDC resistive. Repeatability is ±2% ; accuracy is ±5%. An LED lights when the set point is reached for easy field calibration.