Masterflex Flow Tube for Clip Mount Flow Tube Holder, PFA, 0.5 to 20 LPM; 12 mm Barbed Connection; 10/Bag
Representative image only.
Masterflex Item # EW-24507-35
Build a complete single-use flow sensor system customizable to your application
  • Modular interchangeable flow sensor systems with single-use design
  • Accurate 1% of reading measurement at low flow
  • Low operating cost disposable flow tubes for optimal contamination-free production
  • Ideal for single-use applications while also suitable for long-term measurement
  • Flow tubes and turbine sensors are produced and tested under cleanroom conditions
PRODUCT OPTIONS
$540.00USD / Bag of 10
BUY MORE & SAVE
  • 1-4$540.00USD / Bag of 10
  • 5-7$486.00USD / Bag of 10
  • 8-9$459.00USD / Bag of 10
  • 10+$432.00USD / Bag of 10
$21.00 - $42.00USD / Each
for an additional
$42.00USD / Each
for an additional
$21.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Repeatability<0.15%
  • Accuracy1% of reading
  • Max Pressure (PSI)217.6
  • Min Flow Rate (LPM)0.5
  • Max Flow Rate (LPM)20
  • Material Flow TubeRuby
  • Material FittingPFA
  • Material FloatRuby
  • Min Operating Temperature (° F)-4
  • Min Operating Temperature (° C)-20
  • Max Operating Temperature (° F)176
  • Max Operating Temperature (° C)80
  • Process Connection12 mm hose barb
  • Fluid TypeConductive or non-conductive fluids with no lenticular impurities
  • Viscosity0.8 to 10 cSt
  • DescriptionFlow Tube for Clip Mount Flow Tube Holder, PFA, 0.5 to 20 LPM; 12 mm Barbed Connection; 10/Bag
Key Features
  • Modular interchangeable flow sensor systems with single-use design
  • Accurate 1% of reading measurement at low flow
  • Low operating cost disposable flow tubes for optimal contamination-free production
  • Ideal for single-use applications while also suitable for long-term measurement
  • Flow tubes and turbine sensors are produced and tested under cleanroom conditions
  • Compatible with clip mount flow sensor 32516-02
More About this Item
The Masterflex® single-use flow sensor system is comprised of four components. Select the flow sensor, flow tubes, barcode scanner, and output that suit your application needs for a customizable approach to single-use flow measurement.
These liquid turbine flow sensors are corrosion resistant for long-term, trouble-free performance in demanding situations. Sensor is positioned near the edges of the rotor blades to provide accurate flow measurements for liquids.
Flow tubes come precalibrated to one point. Each flow tube has an attached label that indicates the lot number, serial number, K-factor, and 2-D bar code with all the calibration data. The barcode can quickly be read by the barcode scanner making the calibration process unnecessary when exchanging a flow tube. Eliminate long downtimes and extra calibration while ensuring an accurate measurement with the barcode scanner.
Calibration certificates are available for the flow tubes. Each flow tube comes calibrated without a certificate with the K-factor to 1 point at 50% of the flow range. The optional calibration certificate for the 1-point calibration can be obtained at any time. Calibration with certificate to K-factor at 5 points of flow range can be ordered at the time flow tubes are ordered.
Flow tube sensor, barcode scanner, output options, and calibration certificates sold separately.