Masterflex Flowmeter Acrylic Tripod Base for 03218-50 and 03218-52
Representative image only.
Masterflex Item # EW-03218-58
Easily mount flowmeters on tripod for easy viewing
  • Constructed of durable acrylic
PRODUCT OPTIONS
$58.25USD / Each
BUY MORE & SAVE
  • 1-3$58.25USD / Each
  • 4-9$52.43USD / Each
  • 10-19$49.51USD / Each
  • 20+$46.60USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Width (")1755
  • Material HousingAcrylic
  • DescriptionTripod Base for 03218-50 and 03218-52
Key Features
  • Constructed of durable acrylic
More About this Item
These tripod bases are ideal for benchtop applications. A tripod base is required to use the multitube frame with the common inlet flow pattern; all other multitube frames are designed for panel and tripod mounting. Tripod bases are made of acrylic and include a spirit leveler.