Masterflex Footswitch, On/Off Operation, Wash-Down; 31-Pin Round, 6-ft Cable
Representative image only.
Masterflex Item # EW-07575-84
Easily control your Masterflex® drive with compatible footswitch
  • Choose footswitch with the correct connection for your Masterflex® pump
  • Connection Type: 31-pin round
PRODUCT OPTIONS
$525.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionWash-Down On/Off Footswitch; 31-Pin Round, 6-ft Cable
Key Features
  • Choose footswitch with the correct connection for your Masterflex® pump
  • Connection Type: 31-pin round
More About this Item
Add convenience to repetitive dispensing with a foot-operated on/off switch. Keep hands free to efficiently move containers during routine fill applications or to keep containers stable during the filling process.