Masterflex Gamma Irradiated Sampling Bottle Assembly, Microbially Controlled, PETG Bottle; 2 L

Representative image only.
Masterflex Item # EW-95644-28
Safely transfer, sample, and store biopharmaceutical products and reagents
  • Secure, single-piece cap/tubing assembly eliminates contamination and product loss due to loose connections
  • Assemblies are gamma irradiated—eliminates labor and validation costs associated with assembly and cleaning
  • Designed with 2 L 2000 mL

PRODUCT OPTIONS

Free Certificate of Comliance

$140.00USD / Each

BUY MORE & SAVE

  • 1-4$140.00USD / Each
  • 5-9$126.00USD / Each
  • 10+$112.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Capacity (mL)2000
  • Capacity (L)2
  • Capacity (oz)64
  • MaterialPolyethylene Terephthalate (PET)
  • Cap / Stopper Size53 mm
  • Cap ColorWhite
  • Bottle ColorClear
  • Special FeaturesGamma Irradiated for Microbial Control
  • Tubing ID1/8" ID x 1/4" OD
  • DescriptionGamma Irradiated Sampling Bottle Assembly, Microbially Controlled, PETG Bottle; 2 L
Certificate of Compliance

Includes

Clear PETG square bottle, 53-mm cap, Puri-Flex™ tube/linear assembly with 1/8" ID x 1/4" OD tubing, and 0.2-micron PVDF filter

Key Features

  • Secure, single-piece cap/tubing assembly eliminates contamination and product loss due to loose connections
  • Assemblies are gamma irradiated—eliminates labor and validation costs associated with assembly and cleaning
  • Designed with 2 L 2000 mL

More About this Item

Cap assembly includes a seamless liner with premolded TPE fill and vent tubes to create an unobstructed fluid path. The 24” (61 cm) fill tube is plugged at the terminal end to maintain assembly microbial control until use. Vent tube includes a 25-mm diameter, 0.2-micron PVDF disc filter to prevent contamination. The vent filter is hydrophobic to offer protection loss of product during filling or emptying. The one-piece liner eliminates the risk of bacterial entrapment and contamination and forms an elastomeric seal between the cap and bottle rim.

Puri-Flex™ biopharma tubing meets USP Class VI, FDA 21 CFR 177.2600, and ISO 10993 Part (S) 4/5 requirements. This sealable/weldable TPE tubing has excellent gas barrier properties, low permeability, and is resistant to acids and alkaline. The smooth inner surface reduces platelet adhesion and protein binding.

Bottle assemblies are assembled in an ISO Class 7 cleanroom and are gamma irradiated up to 40 kGy. Each assembly is individually double bagged with a cleanroom label and gamma dot. This gamma-irradiated sampling bottle assemblies protects your sensitive processes and products from accidental contamination while saving time! Gamma-irradiated assemblies help you avoid costly delays that can be associated with outsourced gamma irradiation services. Assemblies are gamma-irradiated to offer optimal microbial control in a critical fluid pathway.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat