Masterflex High-Resolution Brass Metering Valve, straight flow, 12 mL/min
Representative image only.
Masterflex Item # EW-03214-82
Choose from different metal materials based on your application
  • Brass and 316 Stainless Steel material options available
PRODUCT OPTIONS
$94.00USD / Each
BUY MORE & SAVE
  • 1-3$94.00USD / Each
  • 4-9$84.60USD / Each
  • 10-19$79.90USD / Each
  • 20+$75.20USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Port Size (NPT(F))0.125
  • Outlet Port1/8" NPT(F)
  • Connection TypeNPT(F)
  • Inlet Port1/8" NPT(F)
  • Orifice (in)0.042
  • Orifice (mm)1.0668
  • Max Flow Rate For Air (mL/min)400
  • Max Flow Rate For Water (mL/min)12
  • Max Temperature (° F)180
  • Max Temperature (° C)81
  • Wetted MaterialsBrass
  • Flow PatternStraight
  • DescriptionHigh-Resolution Brass Metering Valve, straight flow, 12 mL/min
Key Features
  • Brass and 316 Stainless Steel material options available
More About this Item
Brass valves feature Buna N O-rings; stainless steel valves have Viton® O-rings.