Masterflex I/P® Variable-Speed Air-Powered Drive, 100 to 650 rpm

Representative image only.
Masterflex Item # EW-07589-30
Pump safely in hazardous
environments
  • Flow rate 0.6 to 13 LPM I/P tubing—flow rate depends on drive rpm, pump head, and tubing size
  • Easily adjust motor speed by using included regulator
  • Effortlessly connect to your existing air supply or compressor
  • The 3/4-hp air-powered motor starts smooth, runs cool, and requires little maintenance
  • ATEX Zone 2 approved; rated to EEXII 3 G c IIC T6; NEC Class 1, Division 2, Groups A, B, C, D, T6

SPECS & DESCRIPTION

Specifications & Description

  • Pump SeriesI/P
  • Control TypeAnalog Variable Speed
  • Min RPM (rpm)100
  • Max RPM (rpm)650
  • SpeedVariable
  • Speed Control±10%
  • Max Flow Rate (LPM)17
  • Min Flow Rate (LPM)0.6
  • Max Flow Rate (mL/min)17000
  • Min Flow Rate (mL/min)600
  • Reversible MotorNo
  • No. of Pump Heads Accepted2
  • Type of Pump Heads AcceptedStandard, Easy-Load, High-Performance
  • Motor (watts)560
  • Drive IP ratingIP 34
  • Drive Length (in)11.125
  • Drive Width (in)8.5
  • Drive Height (in)9
  • Drive Length (mm)282
  • Drive Width (mm)216
  • Drive Height (mm)229
  • Open-Head SensorNo
  • Housing MaterialPainted Steel
  • Motor hp3/4
  • DescriptionI/P variable-speed air-powered drive.

Includes

regulator with 5-µm air filter and 1/4'' NPT(F) connection, automatic lubricator, 0 to 100 psi pressure gauge, and muffler.

Key Features

  • Flow rate 0.6 to 13 LPM I/P tubing—flow rate depends on drive rpm, pump head, and tubing size
  • Easily adjust motor speed by using included regulator
  • Effortlessly connect to your existing air supply or compressor
  • The 3/4-hp air-powered motor starts smooth, runs cool, and requires little maintenance
  • ATEX Zone 2 approved; rated to EEXII 3 G c IIC T6; NEC Class 1, Division 2, Groups A, B, C, D, T6

More About this Item

Use this variable-speed air-powered drive when electrical power may be unsafe or impractical. For safe operation, carefully ground to protect from static electricity.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat